Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ KingFisher Plastic Consumables for SARS-CoV-2 Testing

Catalog No. p-9034002
Click to view available options
For Use With (Equipment):
Duo Prime, Flex, Apex, Presto
KingFisher Flex and Presto
Quantity:
100/Pk.
50/Pk.
Includes:
KF 96 DW plate, v-bottom
KingFisher DW 96 Tip Comb

These KingFisher compatible plastic consumables are offered specifically for labs performing COVID-19 testing with the MagMAX Viral/Pathogen Nucleic Acid Purification Kit and the TaqPath COVID-19 Combo Kit.

Optimize performance using Thermo Scientific KingFisher Systems Consumables, designed specifically for use for COVID-19 testing with these instruments. KingFisher instruments utilize disposable plastics made of polypropylene. The plastics, tip combs and microplates, are ideal for magnetic particle processing because of their low binding affinity for biomolecules. The special design enables excellent recovery of magnetic beads.  Specifically tested with  the TaqPath COVID-19 Combo Kit and MagMAX Viral/Pathogen Nucleic Acid Isolation Kit.

For TaqPath COVID-19 testing with MagMAX Viral/Pathogen Nucleic Acid Isolation Kit

TRUSTED_SUSTAINABILITY

Specifications

Format 96 Deep-well Plate
Capacity 96 Samples
For Use With (Equipment) Duo Prime, Flex, Apex, Presto
Includes KF 96 DW plate, v-bottom
Quantity 50/Pk.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.