Learn More
Thermo Scientific™ KingFisher™ Flex Deep Well heating block

Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Format | 96 Deep-well Plate |
| Capacity | 96 Samples |
| For Use With (Equipment) | KingFisher™ Flex |
| Product Line | KingFisher™ |
| Quantity | Each |
| Shipping Condition | Room Temperature |
Safety and Handling
For research use only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.