Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kininogen 1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298281AF488
Description
Kininogen 1 Polyclonal specifically detects Kininogen 1 in Mouse samples. It is validated for ELISA.Specifications
Kininogen 1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
3827 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 488 | |
Alpha-2-thiol proteinase inhibitor, BDK, BK, bradykinin, Fitzgerald factor, High molecular weight kininogen, HMWK, kininogen-1, Williams-Fitzgerald-Flaujeac factor | |
Produced in rabbits immunized with purified, recombinant Mouse Kininogen 1 (NP_001095882.1; Glu 21-Ser 480). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction