Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kininogen 1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299365APC
Description
Kininogen 1 Polyclonal antibody specifically detects Kininogen 1 in Human samples. It is validated for ELISASpecifications
Kininogen 1 | |
Polyclonal | |
ELISA | |
Alpha-2-thiol proteinase inhibitor, BDK, BK, bradykinin, Fitzgerald factor, High molecular weight kininogen, HMWK, kininogen-1, Williams-Fitzgerald-Flaujeac factor | |
Produced in rabbits immunized with purified, recombinant Human Kininogen 1 (Accession#: NP_001095886.1; Gln19-Ser644) | |
0.1 mL | |
Apoptosis, Immunology | |
3827 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction