Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kininogen 1 Antibody (SR2002), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322001100UL
Description
Kininogen 1 Monoclonal antibody specifically detects Kininogen 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
Kininogen 1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
SR2002 | |
Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
Alpha-2-thiol proteinase inhibitor, BDK, BK, bradykinin, Fitzgerald factor, High molecular weight kininogen, HMWK, kininogen-1, Williams-Fitzgerald-Flaujeac factor | |
A synthesized peptide derived from human Kininogen 1 (Uniprot #: P01042) | |
100 μg | |
Apoptosis, Immunology | |
3827 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction