Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kininogen-1 Armenian Hamster anti-Human, Clone: 2B7, MilliporeSigma™

Catalog No. MABN2630100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2630100 100 μg
MABN263025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN2630100 Supplier MilliporeSigma Supplier No. MABN2630100UG
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Kininogen-1 Monoclonal antibody specifically detects Kininogen-1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kininogen-1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 2B7
Conjugate Unconjugated
Formulation Purified armenian Hamster monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Armenian Hamster
Immunogen KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of human Kininogen-1.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.