Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kininogen-1, cleaved form Mouse anti-Human, Clone: 4B12, MilliporeSigma™

Mouse Monoclonal Antibody

$191.34 - $481.79

Specifications

Antigen Kininogen-1, cleaved form
Clone 4B12
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN261925
View Documents
MilliporeSigma
MABN261925UL
25 μL
Each for $191.34
Only null left
Add to Cart
 
MABN2619100
View Documents
MilliporeSigma
MABN2619100UL
100 μL
Each for $481.79
Only null left
Add to Cart
 
Description

Description

Kininogen-1, cleaved form Monoclonal antibody specifically detects Kininogen-1, cleaved form in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation.
Specifications

Specifications

Kininogen-1, cleaved form
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Mouse
Human
KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of human Kininogen-1.
Protein G purified
4B12
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Primary
Stable for 1 year at 2-8°C from date of receipt.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.