Learn More
Invitrogen™ Kininogen 1 Monoclonal Antibody (288422)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA523988
Description
In Western blots, reacts with the full length mouse Kininogen (aa 21-661), but not with the light chain alone. In Western blots, no cross-reactivity with recombinant human (rh) Cystatin D, F, SA, SN, recombinant mouse (rm) Cystatin A, B, C, E/M, rmCystatin/Stefin Homolog, rmFetuin A, rhFetuin B, rmHPRG, rhKininogen, or rhKininostatin is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
This gene uses alternative splicing to generate two different proteins- high molecular weight kininogen (HMWK) and low molecular weight kininogen (LMWK). HMWK is essential for blood coagulation and assembly of the kallikrein-kinin system. Also, bradykinin, a peptide causing numerous physiological effects, is released from HMWK. In contrast to HMWK, LMWK is not involved in blood coagulation. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
Kininogen 1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O08677 | |
KNG1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Kininogen Glu21-Ser661. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Immunoprecipitation, Western Blot | |
288422 | |
Unconjugated | |
KNG1 | |
alpha-1-MAP; Alpha-2-thiol proteinase inhibitor; BDK; BK; Bradykinin; fitzgerald factor; high molecular weight kininogen; H-kininigen; HMWK; Ile-Ser-Bradykinin; Kallidin I; Kallidin II; kininogen; kininogen 1; kininogen 1-like 1; kininogen-1; Kininogen-1 heavy chain; Kininogen-1 light chain; Kng; Kng1; Kng1l1; L-kininogen; low molecular weight (LMW) T-kininogen II; Low molecular weight growth-promoting factor; Lysyl-bradykinin; Major acute phase protein; major acute phase protein (alpha-1-MAP); Thiostatin; T-kinin; T-kininogen 2; T-kininogen 2 heavy chain; T-kininogen 2 light chain; T-kininogen II; T-kininogen II heavy chain; T-kininogen II light chain; williams-Fitzgerald-Flaujeac factor | |
Rat | |
Protein A/G | |
RUO | |
16644 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.