Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kininogen 1 Mouse anti-Human, Clone: 10E8D11, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Kininogen 1 |
---|---|
Clone | 10E8D11 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6559
![]() ![]() |
Proteintech Group Inc
661231IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene uses alternative splicing to generate two different proteins- high molecular weight kininogen (HMWK) and low molecular weight kininogen (LMWK). HMWK is essential for blood coagulation and assembly of the kallikrein-kinin system. Also, bradykinin, a peptide causing numerous physiological effects, is released from HMWK. In contrast to HMWK, LMWK is not involved in blood coagulation. Three transcript variants encoding different isoforms have been found for this gene.Specifications
Kininogen 1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BDK, Fitzgerald factor, HMWK, Ile Ser Bradykinin Bradykinin, Kallidin I Lysyl bradykinin, kininogen 1, KNG, KNG1 | |
KNG1 | |
IgG1 | |
Protein G | |
Antibody |
10E8D11 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P01042 | |
3827 | |
Kininogen 1 Fusion Protein Ag19233 | |
Primary | |
-20°C | |
KNG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title