Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kininogen 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581957
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581957 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581957 Supplier Invitrogen™ Supplier No. PA581957
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALKKYNSQNQ SNNQFVLYRI TEATKTVGSD TFYSFKYEIK EGDCPVQSGK TWQDCEYKDA AKAATGECTA TVGKRSSTKF SVATQTCQIT PAEGPVVTAQ YDCLGCVHPI STQSPDLEPI LRHGIQYFNN N Highest antigen sequence indentity to the following orthologs: Mouse - 62%, Rat - 61%.

This gene uses alternative splicing to generate two different proteins- high molecular weight kininogen (HMWK) and low molecular weight kininogen (LMWK). HMWK is essential for blood coagulation and assembly of the kallikrein-kinin system. Also, bradykinin, a peptide causing numerous physiological effects, is released from HMWK. In contrast to HMWK, LMWK is not involved in blood coagulation. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kininogen 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KNG1
Gene Accession No. P01042
Gene Alias alpha-1-MAP; Alpha-2-thiol proteinase inhibitor; BDK; BK; Bradykinin; fitzgerald factor; high molecular weight kininogen; H-kininigen; HMWK; Ile-Ser-Bradykinin; Kallidin I; Kallidin II; kininogen; kininogen 1; kininogen 1-like 1; kininogen-1; Kininogen-1 heavy chain; Kininogen-1 light chain; Kng; Kng1; Kng1l1; L-kininogen; low molecular weight (LMW) T-kininogen II; Low molecular weight growth-promoting factor; Lysyl-bradykinin; Major acute phase protein; major acute phase protein (alpha-1-MAP); Thiostatin; T-kinin; T-kininogen 2; T-kininogen 2 heavy chain; T-kininogen 2 light chain; T-kininogen II; T-kininogen II heavy chain; T-kininogen II light chain; williams-Fitzgerald-Flaujeac factor
Gene Symbols KNG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KNG1. Recombinant protein control fragment (Product #RP-100525).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3827
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.