Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kininogen 1 Recombinant Rabbit Monoclonal Antibody (JE43-72)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA534837
Description
Positive Control: Human liver tissue, human kidney tissue, mouse stomach lysate.
This gene uses alternative splicing to generate two different proteins- high molecular weight kininogen (HMWK) and low molecular weight kininogen (LMWK). HMWK is essential for blood coagulation and assembly of the kallikrein-kinin system. Also, bradykinin, a peptide causing numerous physiological effects, is released from HMWK. In contrast to HMWK, LMWK is not involved in blood coagulation. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| Kininogen 1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P01042 | |
| KNG1 | |
| Recombinant protein within human kininogen 1 aa 594-644. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JE43-72 | |
| Unconjugated | |
| KNG1 | |
| alpha-1-MAP; Alpha-2-thiol proteinase inhibitor; BDK; BK; Bradykinin; fitzgerald factor; high molecular weight kininogen; H-kininigen; HMWK; Ile-Ser-Bradykinin; Kallidin I; Kallidin II; kininogen; kininogen 1; kininogen 1-like 1; kininogen-1; Kininogen-1 heavy chain; Kininogen-1 light chain; Kng; Kng1; Kng1l1; L-kininogen; low molecular weight (LMW) T-kininogen II; Low molecular weight growth-promoting factor; Lysyl-bradykinin; Major acute phase protein; major acute phase protein (alpha-1-MAP); Thiostatin; T-kinin; T-kininogen 2; T-kininogen 2 heavy chain; T-kininogen 2 light chain; T-kininogen II; T-kininogen II heavy chain; T-kininogen II light chain; williams-Fitzgerald-Flaujeac factor | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3827 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction