Learn More
Invitrogen™ KIP2 Polyclonal Antibody

Description
The antibody detects endogenous levels of total KIP2 protein.
Specifications
Specifications
| Antigen | KIP2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CIB2 |
| Gene Accession No. | O75838, Q9Z309 |
| Gene Alias | 2810434I23Rik; AI449053; calcium and integrin binding family member 2; calcium and integrin-binding family member 2; calcium binding protein Kip 2; calcium binding protein Kip2; CIB2; DFNB48; DNA-dependent protein kinase catalytic subunit-interacting protein 2; kinase interacting protein 2; kinase-interacting protein 2; KIP 2; Kip2; USH1J |
| Gene Symbols | CIB2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.