Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kir2.1 Antibody (S112), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212900C
Description
Kir2.1 Monoclonal antibody specifically detects Kir2.1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Kir2.1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Monkey | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
S112 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Cardiac inward rectifier potassium channel, HHBIRK1, HHIRK1, hIRK1, Inward rectifier K(+) channel Kir2.1, inward rectifier K+ channel KIR2.1, inward rectifier potassium channel 2, IRK-1, IRK1LQT7, Kir2.1, Potassium channel, inwardly rectifying subfamily J member 2, potassium inwardly-rectifying channel, subfamily J, member 2, SQT3 | |
Fusion protein amino acids 41-64 and 189-428 of mouse Kir2.1 | |
0.1 mL | |
Neuroscience | |
3759 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction