Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kir2.1 (KCNJ2) Monoclonal Antibody (S112), APC
GREENER_CHOICE

Catalog No. PIMA545415
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545415 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545415 Supplier Invitrogen™ Supplier No. MA545415
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45415 was sufficient for detection of Kir2.1 in 10 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 45kDa. No cross-reactivity against Kir2.2 or Kir2.3.

The KIR2 subunit family includes 2. 1, 2. 2, 2. 3 and 2. 4. Unlike G protein-coupled KIR3 subunits, KIR2. 1 requires both phosphorylation by PKA and ATP hydrolysis for functional activity. KIR2. 1 is expressed in the superior and inferior collicula and the pontine region of the brain, where it moderates synaptic transmission, like many other potassium channels. In the placenta, KIR2. 1 is expressed throughout gestation in cytotrophoblast cells. In the kidney, KIR2. 1 colocalizes with KIR5. 1 in the proximal tubule. KIR2. 1, 2. 2 and 2. 3 associate with the membrane-associated guanylate kinase synapseassociated protein 97 in the cerebellum and heart. Phosphorylation of KIR2. 2 by protein kinase A inhibits the associates with SAP97.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kir2.1 (KCNJ2)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Microarray
Classification Monoclonal
Clone S112
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4
Gene Kcnj2
Gene Accession No. P35561, P63252, Q64273
Gene Alias ATFB9; cardiac inward rectifier KIR2.1; cardiac inward rectifier potassium channel; EGK_08925; HHBIRK1; HHIRK 1; HHIRK1; HIRK 1; hIRK1; Inward rectifier K; inward rectifier K(+) channel Kir2.1; inward rectifier K+ channel KIR2.1; inward rectifier potassium channel; inward rectifier potassium channel 2; inwardly rectifying potassium channel Kir2.1; IRK 1; IRK1; IRK-1; IRK2; Kcnf1; KCNJ2; KIR2.1; LQT 7; LQT7; Potassium channel, inwardly rectifying subfamily J member 2; potassium channel, inwardly rectifying subfamily J, member 2; potassium inwardly rectifying channel subfamily J member 2; potassium inwardly-rectifying channel, subfamily J, member 2; potassium voltage-gated channel subfamily J member 2; RBL-IRK1; SQT 3; SQT3
Gene Symbols Kcnj2
Host Species Mouse
Immunogen Fusion protein amino acids 41-64 and 189-428 of mouse Kir2.1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16518, 29712, 3759
Target Species Human, Mouse, Rat, Monkey
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.