Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kir2.1 (KCNJ2) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577591
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
The KIR2 subunit family includes 2.1, 2.2, 2.3 and 2.4. Unlike G protein-coupled KIR3 subunits, KIR2.1 requires both phosphorylation by PKA and ATP hydrolysis for functional activity. KIR2.1 is expressed in the superior and inferior collicula and the pontine region of the brain, where it moderates synaptic transmission, like many other potassium channels. In the placenta, KIR2.1 is expressed throughout gestation in cytotrophoblast cells. In the kidney, KIR2.1 colocalizes with KIR5.1 in the proximal tubule. KIR2.1, 2.2 and 2.3 associate with the membrane-associated guanylate kinase synapseassociated protein 97 in the cerebellum and heart. Phosphorylation of KIR2.2 by protein kinase A inhibits the associates with SAP97.
Specifications
Kir2.1 (KCNJ2) | |
Polyclonal | |
Unconjugated | |
KCNJ2 | |
IRK1,Kcnj2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16518, 29712, 3759 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
0.6 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P35561, P63252, Q64273 | |
KCNJ2 | |
Peptide (C)NGVPESTSTDTPPDIDLHN, corresponding to amino acid residues 392-410 of human Kir2.1 | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction