Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kir2.3 (KCNJ4) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102023
Description
Antibody detects endogenous levels of total KCNJ4.
Several different potassium channels are known to be involved with electrical signaling in the nervous system. One class is activated by depolarization whereas a second class is not. The latter are referred to as inwardly rectifying K+ channels, and they have a greater tendency to allow potassium to flow into the cell rather than out of it. This asymmetry in potassium ion conductance plays a key role in the excitability of muscle cells and neurons. The protein encoded by this gene is an integral membrane protein and member of the inward rectifier potassium channel family. The encoded protein has a small unitary conductance compared to other members of this protein family. Two transcript variants encoding the same protein have been found for this gene.
Specifications
Kir2.3 (KCNJ4) | |
Polyclonal | |
Unconjugated | |
KCNJ4 | |
BIR11; Brain inwardly rectifying K(+) channel 2; Hippocampal inward rectifier; hippocampal inward rectifier potassium channel; HIR; HIRK2; HRK1; Inward rectifier K(+) channel Kir2.3; inward rectifier potassium channel 4; IRK3; IRK-3; Kcnf2; KCNJ4; Kir 2.3; Kir2.3; MB-IRK3; Potassium channel, inwardly rectifying subfamily J member 4; potassium channel, inwardly rectifying subfamily J, member 4; potassium inwardly-rectifying channel, subfamily J, member 4; potassium voltage-gated channel subfamily J member 4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
116649, 16520, 3761 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P48050, P52189, P52190 | |
KCNJ4 | |
A synthesized peptide derived from human KCNJ4(Accession P48050), corresponding to amino acid residues R252-Q302. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction