Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR2DL1/CD158a Antibody (HP-MA4), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262191APC
Description
KIR2DL1/CD158a Monoclonal specifically detects KIR2DL1/CD158a in Human samples. It is validated for Flow Cytometry.Specifications
KIR2DL1/CD158a | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
KIR2DL1 | |
Human NK cell line LB2. CD158a/g/h | |
0.1 mL | |
Primary | |
The antibody MEM-112 reacts with CD54 (ICAM-1), a 85-110 kDa type I transmembrane glycoprotein (receptor for rhinovirus). The expression of CD54 is upregulated by activation; it is expressed on activated endothelial cells, T lymphocytes, B lymphocytes, monocytes, macrophages, granulocytes and dendritic cells. | |
Store at 4°C in the dark. |
Flow Cytometry | |
HP-MA4 | |
Flow Cytometry | |
CD158 antigen-like family member A, CD158a antigen, CD158Ankat1, cl-42, killer cell immunoglobulin-like receptor 2DL1, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1, killer Ig receptor, killer inhibitory receptor 2-2-1, KIR221, MHC class I NK cell receptor, Natural killer-associated transcript 1, NKAT, NKAT-1, NKAT1KIR-K64, p58 killer cell inhibitory receptor KIR-K64, p58 natural killer cell receptor clones CL-42/47.11, p58 NK cell inhibitory receptor NKR-K6,47.11, p58 NK receptor CL-42/47.11, p58.1, p58.1 MHC class-I-specific NK receptor | |
Mouse | |
Protein A purified | |
Immunology, Innate Immunity, Signal Transduction | |
3802 | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction