Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                KIR2DL1/CD158a Antibody (MM0439-8J29) - Azide and BSA Free, Novus Biologicals™
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Mouse Monoclonal Antibody
$208.00
Specifications
| Antigen | KIR2DL1/CD158a | 
|---|---|
| Clone | MM0439-8J29 | 
| Concentration | LYOPH | 
| Applications | CyTOF, Flow Cytometry | 
| Classification | Monoclonal | 
Description
KIR2DL1/CD158a Monoclonal specifically detects KIR2DL1/CD158a in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| KIR2DL1/CD158a | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| ADI59742.1 | |
| 3802 | |
| IgG1 | |
| Protein G purified | |
| This antibody only detects KIR2DL1 there is no cross reactivity with other KIRs. | 
| MM0439-8J29 | |
| CyTOF, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD158 antigen-like family member A, CD158a antigen, CD158Ankat1, cl-42, killer cell immunoglobulin-like receptor 2DL1, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1, killer Ig receptor, killer inhibitory receptor 2-2-1, KIR221, MHC class I NK cell receptor, Natural killer-associated transcript 1, NKAT, NKAT-1, NKAT1KIR-K64, p58 killer cell inhibitory receptor KIR-K64, p58 natural killer cell receptor clones CL-42/47.11, p58 NK cell inhibitory receptor NKR-K6,47.11, p58 NK receptor CL-42/47.11, p58.1, p58.1 MHC class-I-specific NK receptor | |
| KIR2DL1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | 
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title