Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR2DL1/CD158a Antibody (MM0439-8J29), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211759PCP
Description
KIR2DL1/CD158a Monoclonal specifically detects KIR2DL1/CD158a in Human samples. It is validated for Flow Cytometry.Specifications
KIR2DL1/CD158a | |
Monoclonal | |
PerCP | |
ADI59742.1 | |
KIR2DL1 | |
Human recombinant KIR2DL1/CD158a | |
0.1 mL | |
Immunology, Innate Immunity, Signal Transduction | |
3802 | |
Human | |
Purified |
Flow Cytometry | |
MM0439-8J29 | |
Flow Cytometry | |
CD158 antigen-like family member A, CD158a antigen, CD158Ankat1, cl-42, killer cell immunoglobulin-like receptor 2DL1, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1, killer Ig receptor, killer inhibitory receptor 2-2-1, KIR221, MHC class I NK cell receptor, Natural killer-associated transcript 1, NKAT, NKAT-1, NKAT1KIR-K64, p58 killer cell inhibitory receptor KIR-K64, p58 natural killer cell receptor clones CL-42/47.11, p58 NK cell inhibitory receptor NKR-K6,47.11, p58 NK receptor CL-42/47.11, p58.1, p58.1 MHC class-I-specific NK receptor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This antibody only detects KIR2DL1 there is no cross reactivity with other KIRs. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction