Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR2DL1/KIR2DS5 Antibody (124), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290181AF700
Description
KIR2DL1/KIR2DS5 Monoclonal specifically detects KIR2DL1/KIR2DS5 in Human samples. It is validated for Flow Cytometry.Specifications
KIR2DL1/KIR2DS5 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
3802 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry | |
124 | |
Flow Cytometry | |
CD158a | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human KIR2DL1/KIR2DS5 (NP_055033.2; Met1-His245). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction