Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR2DL1/KIR2DS5 Antibody (143211), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1844Z
Description
KIR2DL1/KIR2DS5 Monoclonal antibody specifically detects KIR2DL1/KIR2DS5 in Human samples. It is validated for Flow Cytometry, CyTOF-reportedSpecifications
KIR2DL1/KIR2DS5 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3802 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, CyTOF | |
143211 | |
Flow Cytometry, CyTOF-reported | |
CD158a | |
BaF3 mouse pro-B cell line transfected with human KIR2DL1/KIR2DS5, Accession # P43626 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction