Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIR2DL1 Monoclonal Antibody (07)
GREENER_CHOICE

Catalog No. PIMA530607
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530607 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530607 Supplier Invitrogen™ Supplier No. MA530607
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human KIR2DL1/CD158a.

This gene encodes a myotubularin-related protein. The encoded protein is a phosphoinositide phosphatase that specifically dephosphorylates phosphatidylinositol 3,5-biphosphate and phosphatidylinositol 3-phosphate. Mutations in this gene are correlated with autosomal dominant centronuclear myopathy. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR2DL1
Applications ELISA
Classification Monoclonal
Clone 07
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene KIR2DL1
Gene Accession No. P43626
Gene Alias CD158 antigen-like family member A; CD158a; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1; killer cell immunoglobulin-like receptor 2DL1; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1; killer Ig receptor; killer inhibitory receptor 2-2-1; KIR221; KIR2DL1; KIR2DS1; KIR-K64; MHC class I NK cell receptor; natural killer cell receptor; Natural killer-associated transcript 1; NKAT; NKAT1; NKAT-1; p58 killer cell inhibitory receptor KIR-K64; p58 natural killer cell receptor clones CL-42/47.11; p58 NK cell inhibitory receptor NKR-K6; p58 NK receptor CL-42/47.11; p58.1; p58.1 MHC class-I-specific NK receptor
Gene Symbols KIR2DL1
Host Species Mouse
Immunogen Recombinant Human KIR2DL1/CD158a Protein (Met1-His245).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3802
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.