Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIR2DL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114214
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114214 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114214 Supplier Invitrogen™ Supplier No. PA5114214
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DEQDPQEVTY TQLNHCVFTQ RKITRPSQRP KTPPTDIIVY TELPNAESRS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

This gene encodes a myotubularin-related protein. The encoded protein is a phosphoinositide phosphatase that specifically dephosphorylates phosphatidylinositol 3,5-biphosphate and phosphatidylinositol 3-phosphate. Mutations in this gene are correlated with autosomal dominant centronuclear myopathy. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR2DL1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KIR2DL1
Gene Accession No. P43626
Gene Alias CD158 antigen-like family member A; CD158a; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1; killer cell immunoglobulin-like receptor 2DL1; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1; killer Ig receptor; killer inhibitory receptor 2-2-1; KIR221; KIR2DL1; KIR2DS1; KIR-K64; MHC class I NK cell receptor; natural killer cell receptor; Natural killer-associated transcript 1; NKAT; NKAT1; NKAT-1; p58 killer cell inhibitory receptor KIR-K64; p58 natural killer cell receptor clones CL-42/47.11; p58 NK cell inhibitory receptor NKR-K6; p58 NK receptor CL-42/47.11; p58.1; p58.1 MHC class-I-specific NK receptor
Gene Symbols KIR2DL1
Host Species Rabbit
Immunogen Immunogen sequence: DEQDPQEVTY TQLNHCVFTQ RKITRPSQRP KTPPTDIIVY TELPNAESRS (aa 293-342).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3802
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.