Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR2DL1/S1/S3/S5 (CD158) Mouse anti-Human, Alexa Fluor 647, Clone: HP-MA4, BD Pharmingen™
Mouse Monoclonal Antibody
$138.00 - $338.00
Specifications
Antigen | KIR2DL1/S1/S3/S5 (CD158) |
---|---|
Clone | HP-MA4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 647 |
Description
KIR2DL1/S1/S3/S5 (CD158) Monoclonal antibody specifically detects KIR2DL1/S1/S3/S5 (CD158) in Human samples. It is validated for Flow CytometrySpecifications
KIR2DL1/S1/S3/S5 (CD158) | |
Flow Cytometry | |
Alexa Fluor 647 | |
Mouse | |
Human | |
KIR2DL1 (CD158a/NKAT-1); KIR2DS1 (CD158h); KIR2DS3 (NKAT-7); KIR2DS5 (CD158g/NKAT-9) | |
IgG2b κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
HP-MA4 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human NK Clone LB2 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title