Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIR2DL3 Monoclonal Antibody (GL183)
GREENER_CHOICE

Catalog No. 01672411
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01672411 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01672411 Supplier Invitrogen™ Supplier No. MA135918
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GL183 recognizes human Killer cell immunoglobulin-like receptor 2DL3, also known as CD158b, KIR-023GB, MHC class I NK cell receptor, p58 natural killer cell receptor clone CL-6 or Natural killer-associated transcript 2. CD158b is a 341 amino acid, ~58 kDa single pass type-1 transmembrane glycoprotein containing two Ig-like C2-type domains. expressed by a subset of NK cells.This antibody also recognizes a ~50 kDa molecule in some NK clones, which is highly homologous to p58.2 in the extracellular domain, but has a shorter cytoplasmic tail (Moretta et al. 1985). Both molecules are members of the newly described natural killer cell receptor family.CD158b functions as a receptor specific for HLA Class I molecules, including Cw3 and related HLA-C alleles. Mouse anti Human CD158b antibody, clone GL183 can restore the lysis by human NK clones of otherwise lysis protected targets expressing Cw3.

NKAT (NK-associated transcripts) gene products, known as killer immuno-globulin-like receptors or KIRs, downregulate the cytotoxicity of NK cells upon recognition of specific class I major histocompatibility complex (MHC) molecules on target cells. This family of receptors is characterized by an extracellular region with two to three immunoglobulin-superfamily domains and a cytoplasmic domain with an antigen receptor activation motif (ARAM). KIRs and other inhibitory receptors also possess a common cytoplasmic sequence (I/VxYxxL/V) known as an ITIM (immunoreceptor tyrosine-based inhibitory motif). The human inhibitory human killer cell immunoglobulin-like receptor 2DL3 (KIR2DL3), also referred to as CD158b, is an inhibitory receptor that is specific for the human MHC class I molecule HLA-Cw3 and related alleles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR2DL3
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone GL183
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene KIR2DL3
Gene Accession No. P43628
Gene Alias CD158 antigen-like family member B2; CD158b; CD158B2; CU464054.1; GL183; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 3; killer cell immunoglobulin-like receptor 2DL3; killer cell immunoglobulin-like receptor two domains long cytoplasmic tail 3; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 3; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5; killer Ig-like inhibitory receptor; killer inhibitory receptor cl 2-3; KIR-023GB; KIR2DL3; KIR2DS3; KIR2DS5; KIRCL23; KIR-K7b; KIR-K7c; MGC129943; MHC class I NK cell receptor; natural killer associated transcript 2; natural killer cell inhibitory receptor KIR2DL3; natural killer cell receptor; natural killer-associated transcript 2; NKAT; NKAT2; NKAT-2; NKAT2A; NKAT2B; NK-receptor; p58; p58 natural killer cell receptor clone CL-6; p58 NK receptor CL-6; p58.2 MHC class-I specific NK receptor; p58.2 MHC class-I-specific NK receptor
Gene Symbols KIR2DL3
Host Species Mouse
Immunogen NK cell clone E57.
Purification Method Purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3804
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.