Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIR2DL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102464
Description
Antibody detects endogenous levels of total KIR2DL3.
NKAT (NK-associated transcripts) gene products, known as killer immuno-globulin-like receptors or KIRs, downregulate the cytotoxicity of NK cells upon recognition of specific class I major histocompatibility complex (MHC) molecules on target cells. This family of receptors is characterized by an extracellular region with two to three immunoglobulin-superfamily domains and a cytoplasmic domain with an antigen receptor activation motif (ARAM). KIRs and other inhibitory receptors also possess a common cytoplasmic sequence (I/VxYxxL/V) known as an ITIM (immunoreceptor tyrosine-based inhibitory motif). The human inhibitory human killer cell immunoglobulin-like receptor 2DL3 (KIR2DL3), also referred to as CD158b, is an inhibitory receptor that is specific for the human MHC class I molecule HLA-Cw3 and related alleles.
Specifications
KIR2DL3 | |
Polyclonal | |
Unconjugated | |
KIR2DL3 | |
CD158 antigen-like family member B2; CD158b; CD158B2; CU464054.1; GL183; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 3; killer cell immunoglobulin-like receptor 2DL3; killer cell immunoglobulin-like receptor two domains long cytoplasmic tail 3; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 3; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5; killer Ig-like inhibitory receptor; killer inhibitory receptor cl 2-3; KIR-023GB; KIR2DL3; KIR2DS3; KIR2DS5; KIRCL23; KIR-K7b; KIR-K7c; MGC129943; MHC class I NK cell receptor; natural killer associated transcript 2; natural killer cell inhibitory receptor KIR2DL3; natural killer cell receptor; natural killer-associated transcript 2; NKAT; NKAT2; NKAT-2; NKAT2A; NKAT2B; NK-receptor; p58; p58 natural killer cell receptor clone CL-6; p58 NK receptor CL-6; p58.2 MHC class-I specific NK receptor; p58.2 MHC class-I-specific NK receptor | |
Rabbit | |
Affinity chromatography | |
RUO | |
3804 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P43628 | |
KIR2DL3 | |
A synthesized peptide derived from human KIR2DL3(Accession P43628), corresponding to amino acid residues R203-V253. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction