Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR2DL4/CD158d Antibody (MM0441-8L21), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211761UV
Description
KIR2DL4/CD158d Monoclonal specifically detects KIR2DL4/CD158d in Human samples. It is validated for Flow Cytometry.Specifications
KIR2DL4/CD158d | |
Monoclonal | |
DyLight 350 | |
103AS, 15.212, CD158 antigen-like family member D, CD158d antigen, CD158DKIR103ASKiller cell inhibitory receptor 103AS, G9P, killer cell immunoglobulin-like receptor 2DL4, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4, killer Ig receptor, KIR103, KIR-103AS, MHC class I NK cell receptor KIR103AS, natural killer cell inhibitory receptor, NK cell receptor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
MM0441-8L21 | |
Flow Cytometry | |
KIR2DL4 | |
Human recombinant KIR2DL4/CD158d | |
0.1 mL | |
Immunology | |
3805 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction