Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR2DL4/CD158d Antibody (MM0441-8L21), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211761JF646
Description
KIR2DL4/CD158d Monoclonal specifically detects KIR2DL4/CD158d in Human samples. It is validated for Flow Cytometry.Specifications
KIR2DL4/CD158d | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KIR2DL4 | |
Human recombinant KIR2DL4/CD158d | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
MM0441-8L21 | |
Flow Cytometry | |
103AS, 15.212, CD158 antigen-like family member D, CD158d antigen, CD158DKIR103ASKiller cell inhibitory receptor 103AS, G9P, killer cell immunoglobulin-like receptor 2DL4, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4, killer Ig receptor, KIR103, KIR-103AS, MHC class I NK cell receptor KIR103AS, natural killer cell inhibitory receptor, NK cell receptor | |
Mouse | |
Protein G purified | |
Immunology | |
3805 | |
Store at 4°C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction