Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIR2DL4 Monoclonal Antibody (11)
GREENER_CHOICE

Catalog No. PIMA530609
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530609 Supplier Invitrogen™ Supplier No. MA530609
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human KIR2DL4/CD158D.

CD158d / KIR2DL4 is a KIR family member that shares structural features with both activating and inhibitory receptors and may mediate different functions under different circumstances. It contains cytoplasmic ITIM, suggesting inhibitory function, but also transmembrane domain similar to those of activating KIRs. It has been reported that CD158d serves as an inhibitory receptor for peripheral and uterine NK cells, but its ligation with soluble mAbs (unlike immobilized mAbs) results in activation of IFN-gamma secretion. CD158d also binds both membrane form and soluble form of its ligand HLA-G.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR2DL4
Applications ELISA
Classification Monoclonal
Clone 11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene KIR2DL4
Gene Accession No. Q99706
Gene Alias CD158 antigen-like family member D; CD158D; G9P; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 4; killer cell immunoglobulin-like receptor 2DL4; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4; killer cell inhibitory receptor 103AS; killer Ig receptor; killer-cell immunoglobulin-like receptor; killer-cell immunoglobulin-like receptor 2DL4; KIR; KIR103; KIR103AS; KIR-103AS; KIR2DL4; KIR-2DL4; MHC class I NK cell receptor KIR103AS; natural killer cell inhibitory receptor; natural killer cell receptor; NK cell receptor
Gene Symbols KIR2DL4
Host Species Mouse
Immunogen Recombinant Human KIR2DL4/CD158D Protein (Met1-His242).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3805
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.