Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIR2DL4 Monoclonal Antibody (mAb#33), Alexa Fluor™ 647
GREENER_CHOICE

Catalog No. PIMA544143
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544143 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544143 Supplier Invitrogen™ Supplier No. MA544143
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody recognizes extracellular portion of CD158d / KIR2DL4, a 45 kDa NK cell marker. Cell surface expression and function of CD158d / KIR2DL4 depends on genotype of particular individuals.

CD158d / KIR2DL4 is a KIR family member that shares structural features with both activating and inhibitory receptors and may mediate different functions under different circumstances. It contains cytoplasmic ITIM, suggesting inhibitory function, but also transmembrane domain similar to those of activating KIRs. It has been reported that CD158d serves as an inhibitory receptor for peripheral and uterine NK cells, but its ligation with soluble mAbs (unlike immobilized mAbs) results in activation of IFN-gamma secretion. CD158d also binds both membrane form and soluble form of its ligand HLA-G.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR2DL4
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone mAb#33
Concentration 0.160 mg/mL
Conjugate Alexa Fluor 647
Formulation PBS with 0.2% BSA and 15mM sodium azide
Gene KIR2DL4
Gene Accession No. Q99706
Gene Alias CD158 antigen-like family member D; CD158D; G9P; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 4; killer cell immunoglobulin-like receptor 2DL4; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4; killer cell inhibitory receptor 103AS; killer Ig receptor; killer-cell immunoglobulin-like receptor; killer-cell immunoglobulin-like receptor 2DL4; KIR; KIR103; KIR103AS; KIR-103AS; KIR2DL4; KIR-2DL4; MHC class I NK cell receptor KIR103AS; natural killer cell inhibitory receptor; natural killer cell receptor; NK cell receptor
Gene Symbols KIR2DL4
Host Species Mouse
Immunogen NK3.3 cells and KIR2DL4-Ig fusion protein.
Purification Method Size-exclusion chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3805
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.