Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR2DS1/CD158h Antibody (1127B), PE/Cy7, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB8887PECY7
Description
KIR2DS1/CD158h Monoclonal antibody specifically detects KIR2DS1/CD158h in Human samples. It is validated for Flow CytometrySpecifications
| KIR2DS1/CD158h | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3806 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG |
| Flow Cytometry | |
| 1127B | |
| Flow Cytometry | |
| killer cell immunoglobulin-like receptor 2DS1, CD158a, CD158H, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 1, p50.1 | |
| HEK293 human embryonic kidney cell line transfected with human KIR2DS1/CD158h, Met1-His245, Accession # Q14954 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction