Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DS2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017413 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017413 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017413 Supplier Novus Biologicals Supplier No. NBP282102
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIR2DS2 Polyclonal specifically detects KIR2DS2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen KIR2DS2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL
Formulation PBS
Gene Alias 183ActI, CD158b, CD158J, cl-49, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 2, NKAT5, p58 KIR
Gene Symbols KIR2DS2
Host Species Rabbit
Immunogen KIR2DS2 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human KIR2DS2. The immunogen is located within the last 50 amino acids of KIR2DS2. Amino Acid Squence: EPAGNRTVNSEDSDEQDH
Molecular Weight of Antigen 34, 42 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100132285
Test Specificity KIR2DS2 antibody is human specific. Multiple isoforms are known to exist.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.