Learn More
Invitrogen™ KIR2DS3 Polyclonal Antibody

Description
KIR2DS3 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | KIR2DS3 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | KIR2DS3 |
| Gene Accession No. | Q14952 |
| Gene Alias | killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 3; killer cell immunoglobulin-like receptor 2DS3; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 3; KIR2DS3; MHC class I NK cell receptor; natural killer cell inhibitory receptor; natural killer-associated transcript 7; NKAT7; NKAT-7 |
| Gene Symbols | KIR2DS3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.