Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kir3.1 (KCNJ3) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597258
Description
Immunogen sequence: NGRCNVQHGN LGSETSRYLS DLFTTLVDLK WRWNLFIFIL TYTVAWLFMA SMWWVIAYTR GDLNKAHVGN YTPCVANVYN FPSAFLFFIE TEATIGYGYR Y; Positive Samples: Mouse brain, Rat brain; Cellular Location: Membrane, Multi-pass membrane protein.
This potassium channel is controlled by G proteins. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. This receptor plays a crucial role in regulating the heartbeat.
Specifications
Kir3.1 (KCNJ3) | |
Polyclonal | |
Unconjugated | |
KCNJ3 | |
EGK_04447; G protein-activated inward rectifier potassium channel 1; GIRK1; GIRK-1; HGIRK1; inward rectifier K(+) channel Kir3.1; inward rectifier K+ channel KIR3.1; Kcnf3; Kcnj3; KGA; KGB1; KIR3.1; potassium channel inwarding rectifying channel subfamily J member 3; potassium channel subunit Kir3.1 type 3 delta; potassium channel, inwardly rectifying subfamily J member 3; potassium channel, inwardly rectifying subfamily J, member 3; potassium inwardly rectifying channel subfamily J member 3; potassium inwardly-rectifying channel subfamily J member 3 splice variant 1e; potassium inwardly-rectifying channel, subfamily J, member 3; potassium voltage-gated channel subfamily J member 3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
16519, 3760, 50599 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.912 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P48549, P63250, P63251 | |
KCNJ3 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human KCNJ3 (NP_0022301). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction