Learn More
Invitrogen™ Kir3.1 (KCNJ3) Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (98%), Rat (98%), Chicken (91%), Rhesus Monkey (100%), Bovine (99%), Guinea pig (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Kir3.1 (KCNJ3) |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | KCNJ3 |
| Gene Accession No. | P48549, P63250 |
| Gene Alias | EGK_04447; G protein-activated inward rectifier potassium channel 1; GIRK1; GIRK-1; HGIRK1; inward rectifier K(+) channel Kir3.1; inward rectifier K+ channel KIR3.1; Kcnf3; Kcnj3; KGA; KGB1; KIR3.1; potassium channel inwarding rectifying channel subfamily J member 3; potassium channel subunit Kir3.1 type 3 delta; potassium channel, inwardly rectifying subfamily J member 3; potassium channel, inwardly rectifying subfamily J, member 3; potassium inwardly rectifying channel subfamily J member 3; potassium inwardly-rectifying channel subfamily J member 3 splice variant 1e; potassium inwardly-rectifying channel, subfamily J, member 3; potassium voltage-gated channel subfamily J member 3 |
| Gene Symbols | KCNJ3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.