Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kir3.2 (KCNJ6) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518802
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518802 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518802 Supplier Invitrogen™ Supplier No. PA518802
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins and may be involved in the regulation of insulin secretion by glucose. It associates with two other G-protein-activated potassium channels to form a heteromultimeric pore-forming complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kir3.2 (KCNJ6)
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene KCNJ6
Gene Accession No. P48051
Gene Alias BIR1; G protein-activated inward rectifier potassium channel 2; Girk2; GIRK-2; hiGIRK2; Inward rectifier K(+) channel Kir3.2; inward rectifier potassium channel KIR3.2; KATP2; KATP-2; KCNJ6; Kcnj7; KIR3.2; KPLBS; MGC126596; Potassium channel, inwardly rectifying subfamily J member 6; potassium channel, inwardly rectifying subfamily J, member 6; potassium inwardly-rectifying channel J6; potassium inwardly-rectifying channel, subfamily J, member 6; potassium voltage-gated channel subfamily J member 6; W; weaver; wv
Gene Symbols KCNJ6
Host Species Goat
Immunogen Synthetic peptide sequence (SSKLNQHAELET) corresponding to the C-terminus amino acids of KCNJ6 (aa 386-398).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3763
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.