Learn More
Invitrogen™ Kir3.2 (KCNJ6) Polyclonal Antibody

Description
PA1-9548 detects KCNJ6 in human samples. PA1-9548 has been successfully used in Western blot procedures. By Western blot, PA1-9548 detects a 49 kDa protein representing KCNJ6. PA1-9548 immunogen is a purified recombinant KCNJ6 corresponding to the chromosome 21 gene (Human).
Specifications
Specifications
| Antigen | Kir3.2 (KCNJ6) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | KCNJ6 |
| Gene Accession No. | P48051 |
| Gene Alias | BIR1; G protein-activated inward rectifier potassium channel 2; Girk2; GIRK-2; hiGIRK2; Inward rectifier K(+) channel Kir3.2; inward rectifier potassium channel KIR3.2; KATP2; KATP-2; KCNJ6; Kcnj7; KIR3.2; KPLBS; MGC126596; Potassium channel, inwardly rectifying subfamily J member 6; potassium channel, inwardly rectifying subfamily J, member 6; potassium inwardly-rectifying channel J6; potassium inwardly-rectifying channel, subfamily J, member 6; potassium voltage-gated channel subfamily J member 6; W; weaver; wv |
| Gene Symbols | KCNJ6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.