Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kir3.3 (KCNJ9) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541029 Supplier Invitrogen™ Supplier No. PA541029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CQARSSYLVD EVLWGHRFTS VLTLEDGFYE VDYASFHETF EVPTPSCSAR Sequence homology: Cow: 100%; Dog: 93%; Goat: 93%; Guinea Pig: 100%; Horse: 77%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 92%.

Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins. It associates with another G-protein-activated potassium channel to form a heteromultimeric pore-forming complex. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kir3.3 (KCNJ9)
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCNJ9
Gene Accession No. Q92806
Gene Alias 1700085N21Rik; G protein-activated inward rectifier potassium channel 3; G protein-coupled inward rectifier potassium channel; Girk3; GIRK-3; Inward rectifier K(+) channel Kir3.3; inwardly rectifier K(+) channel Kir3.3; inwardly rectifier K+ channel KIR3.3; KCNJ9; Kir3.3; mbGIRK3; Potassium channel, inwardly rectifying subfamily J member 9; potassium channel, inwardly rectifying subfamily J, member 9; potassium inwardly rectifying channel subfamily J member 9; potassium inwardly-rectifying channel subfamily J9; potassium inwardly-rectifying channel, subfamily J, member 9; potassium voltage-gated channel subfamily J member 9; si:dkey-42d18.2
Gene Symbols KCNJ9
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human KCNJ9.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3765
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.