Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kir3.4 (KCNJ5) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119662
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119662 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119662 Supplier Invitrogen™ Supplier No. PA5119662
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Mouse heart tissue, rat heart tissue, 293T, HepG2, LOVO, human lung cancer tissue, human pancreas tissue, mouse pancreas tissue. Subcellular Location: Membrane.

G protein-coupled inwardly rectifying potassium channels (Kir3.1 through Kir3.4) are coupled to numerous neurotransmitter receptors in the brain and are abundantly expressed in the olfactory bulb, hippocampus, neocortex, dentate gyrus, cerebellar cortex and thalamus regions of the brain. Also known as GIRK, Kir3 potassium channels localize to the soma and dendrites as well as axons of neurons. Liberated Gbg subunits from G protein heterotrimers bind to and regulate Kir3 channel activity. Gb3- and Gb4-containing Gbg dimers bind directly to cytoplasmic domains of Kir3 proteins and increase the K+ current while Gb5-containing Gbg dimers inhibit Kir3 K+ current. Kir3 activity is also inhibited by tyrosine phosphorylation. Brain-derived neurotrophic factor activates receptor tyrosine kinase B, which then phosphorylates Kir3 tyrosine residues, effectively inactivating the Kir3 channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kir3.4 (KCNJ5)
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene KCNJ5
Gene Accession No. P48544, P48545, P48548
Gene Alias cardiac ATP-sensitive potassium channel; cardiac inward rectifier; CIR; G protein-activated inward rectifier potassium channel 4; GIRK4; GIRK-4; Heart KATP channel; inward rectifier K(+) channel Kir3.4; inward rectifier K+ channel KIR3.4; IRK4; IRK-4; KATP1; KATP-1; KCNJ5; Kir3.4; LQT13; Potassium channel, inwardly rectifying subfamily J member 5; potassium channel, inwardly rectifying subfamily J, member 5; potassium inwardly-rectifying channel, subfamily J, member 5; potassium voltage-gated channel subfamily J member 5
Gene Symbols KCNJ5
Host Species Rabbit
Immunogen Synthetic peptide within N-terminal human Kir3.4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16521, 29713, 3762
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.