Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kir3.4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Kir3.4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Kir3.4 Polyclonal specifically detects Kir3.4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Kir3.4 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
3762 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Cardiac inward rectifier, CIRKIR3.4, G protein-activated inward rectifier potassium channel 4, GIRK-4, GIRK4Kir3.4, Heart KATP channel, Inward rectifier K(+) channel Kir3.4, inward rectifier K+ channel KIR3.4, IRK-4, KATP-1, KATP1cardiac ATP-sensitive potassium channel, LQT13, Potassium channel, inwardly rectifying subfamily J member 5, potassium inwardly-rectifying channel, subfamily J, member 5 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Kir3.4 (NP_000881.3). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title