Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR3DL1 Antibody (177407), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB12251I
This item is not returnable.
View return policy
Description
KIR3DL1 Monoclonal antibody specifically detects KIR3DL1 in Human samples. It is validated for Flow CytometrySpecifications
KIR3DL1 | |
Monoclonal | |
Janelia Fluor 549 | |
AMB11, CD158 antigen-like family member E, CD158E, CD158e antigen, CD158e1, CD158e1/2, CD158e2, cl-11, cl-2, HLA-BW4-specific inhibitory NK cell receptor, killer cell immunoglobulin-like receptor 3DL1, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail1,nkat3, killer Ig receptor, KIR antigen 3DL1, KIR3DL1/S1, KIR3DS1, MGC119726, MGC119728, MGC126589, MGC126591, MHC class I NK cell receptor, Natural killer-associated transcript 3, NKAT-3, NKAT3KIR, NKB1B, NKB1CD158E1, NK-receptor, p70 killer cell inhibitory receptor, p70 natural killer cell receptor clones CL-2/CL-11, p70 NK receptor CL-2/CL-11 | |
BaF3 mouse pro-B cell line transfected with human KIR3DL1, Accession # P43629 | |
0.5 mL | |
Immunology | |
3811 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry | |
177407 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction