Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR3DL1 Antibody (MM0443-1L34) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
| Antigen | KIR3DL1 |
|---|---|
| Clone | MM0443-1L34 |
| Concentration | LYOPH |
| Applications | Blocking Assay, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
Description
KIR3DL1 Monoclonal specifically detects KIR3DL1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready, Block/Neutralize.Specifications
| KIR3DL1 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| ADM64608.1 | |
| 3811 | |
| IgG1 | |
| Protein G purified | |
| This antibody was selected for its ability to specifically detect human KIR3DL1. |
| MM0443-1L34 | |
| Blocking Assay, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| AMB11, CD158 antigen-like family member E, CD158E, CD158e antigen, CD158e1, CD158e1/2, CD158e2, cl-11, cl-2, HLA-BW4-specific inhibitory NK cell receptor, killer cell immunoglobulin-like receptor 3DL1, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail1,nkat3, killer Ig receptor, KIR antigen 3DL1, KIR3DL1/S1, KIR3DS1, MGC119726, MGC119728, MGC126589, MGC126591, MHC class I NK cell receptor, Natural killer-associated transcript 3, NKAT-3, NKAT3KIR, NKB1B, NKB1CD158E1, NK-receptor, p70 killer cell inhibitory receptor, p70 natural killer cell receptor clones CL-2/CL-11, p70 NK receptor CL-2/CL-11 | |
| KIR3DL1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title