Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIR3DL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534816
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534816 Supplier Invitrogen™ Supplier No. PA534816
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. The KIR genes are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC). The gene content of the KIR gene cluster varies among haplotypes, although several 'framework' genes are found in all haplotypes (KIR3DL3, KIR3DP1, KIR3DL4, KIR3DL2). The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain. KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM), while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR3DL2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Kir3dl2
Gene Accession No. P43630
Gene Alias 3DL2; CD158 antigen-like family member K; CD158K; killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 2; killer cell immunoglobulin-like receptor 3DL2; killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 2; killer Ig receptor; killer immunoglobulin-like receptor-like 2; killer-cell immunoglobulin-like receptor; killer-cell immunoglobulin-like receptor 3DL2; KIR antigen 3DL2; KIR3DL2; KIR-3DL2; Kirl2; KIR-like receptor 2; LOC727787; MHC class I NK cell receptor; natural killer cell receptor; natural killer-associated transcript 4; NKAT4; NKAT-4; NKAT4B; p140; p70 killer cell inhibitory receptor; p70 natural killer cell receptor clone CL-5; p70 NK receptor CL-5
Gene Symbols Kir3dl2
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Extracellular domain of human LOC727787. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3812
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.