Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIR3DL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598760
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598760 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598760 Supplier Invitrogen™ Supplier No. PA598760
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIR3DL3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

KIR3DL3 is transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. This protein are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR3DL3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KIR3DL3
Gene Accession No. Q8N743
Gene Alias CD158 antigen-like family member Z; CD158Z; killer cell Ig-like receptor KIR3DL7; killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 3; killer cell immunoglobulin-like receptor 3DL3; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 2; killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 3; killer cell inhibitory receptor 1; killer-cell immunoglobulin-like receptor 3DL3; KIR2DS2; KIR3DL3; KIR3DL7; KIR44; KIRC1; natural killer cell receptor
Gene Symbols KIR3DL3
Host Species Rabbit
Immunogen Recombinant Human Killer cell immunoglobulin-like receptor 3DL3 protein (26-322AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115653
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.