Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR3DS1/CD158e2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | KIR3DS1/CD158e2 |
---|---|
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
KIR3DS1/CD158e2 Polyclonal specifically detects KIR3DS1/CD158e2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KIR3DS1/CD158e2 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
Recombinant fusion protein containing a sequence corresponding to amino acids 80-340 of human KIR3DS1/CD158e2 (NP_001077008.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
3813 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title