Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIR3DS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549621
Description
KIR3DS1 Polyclonal Antibody for Western Blot, Flow
The killer immunoglobulin-like receptors (KIRs) on natural killer (NK) cells regulate the inhibition and activation of NK-cell responses through recognition of human leukocyte antigen (HLA) class I molecules. KIR3DL1, a receptor for HLA-B antigens with the Bw4 allele, transmits an inhibitory signal to prevent killer cell-mediated cytoxicity. KIR3DL1 encodes a 444 amino acid type I transmembrane protein, containing three immunoglobulin-like C2-type domains.
Specifications
KIR3DS1 | |
Polyclonal | |
Unconjugated | |
KIR3DS1 | |
CD158E2; killer cell immunoglobulin like receptor, three Ig domains and short cytoplasmic tail 1; killer cell immunoglobulin-like receptor 3DS1; killer cell immunoglobulin-like receptor KIR3DS1; killer cell immunoglobulin-like receptor, three domains, short cytoplasmic tail, 1; killer Ig receptor; KIR-123FM; KIR3DS1; KIR-G1; MHC class I NK cell receptor; natural killer cell receptor; natural killer-associated transcript 10; NKAT10; NKAT-10 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
3813 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q14943 | |
KIR3DS1 | |
KLH conjugated synthetic peptide between 361-387 amino acids from the C-terminal region of human KIR3DS1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction