Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIR3DS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597295
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597295 Supplier Invitrogen™ Supplier No. PA597295
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FNMSPVTTAH AGNYTCRGSH PHSPTGWSAP SNPMVIMVTG NHRKPSLLAH PGPLVKSGER VILQCWSDIM FEHFFLHKEW ISKDPSRLVG QIHDGVSKAN FSIGSMMRAL AGTYRCYGSV THTPYQLSAP SDPLDIVVTG LYEKPSLSAQ PGPKVQAGES VTLSCSSRSS YDMYHLSREG GAHERRLPAV RKVNRTFQAD FPLGPATHGG TYRCFGSFRH SPYEWSDPSD PLLVSVTGNP SSSWPSPTEP SSKSGNLRHL H; Positive Samples: Jurkat, A-549, THP-1, BT-474, B-cell, Mouse liver, Mouse kidney, Mouse pancreas, Rat liver; Cellular Location: Cell membrane, Single-pass type I membrane protein.

The killer immunoglobulin-like receptors (KIRs) on natural killer (NK) cells regulate the inhibition and activation of NK-cell responses through recognition of human leukocyte antigen (HLA) class I molecules. KIR3DL1, a receptor for HLA-B antigens with the Bw4 allele, transmits an inhibitory signal to prevent killer cell-mediated cytoxicity. KIR3DL1 encodes a 444 amino acid type I transmembrane protein, containing three immunoglobulin-like C2-type domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR3DS1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KIR3DS1
Gene Accession No. Q14943
Gene Alias CD158E2; killer cell immunoglobulin like receptor, three Ig domains and short cytoplasmic tail 1; killer cell immunoglobulin-like receptor 3DS1; killer cell immunoglobulin-like receptor KIR3DS1; killer cell immunoglobulin-like receptor, three domains, short cytoplasmic tail, 1; killer Ig receptor; KIR-123FM; KIR3DS1; KIR-G1; MHC class I NK cell receptor; natural killer cell receptor; natural killer-associated transcript 10; NKAT10; NKAT-10
Gene Symbols KIR3DS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 80-340 of human KIR3DS1 (NP_0010770081).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3813
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.