Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kir4.1 (KCNJ10) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585057
Description
Immunogen sequence: EFTPAISLSA SGKYIADFSL FDQVVKVASP SGLRDSTVRY GDPEKLKLEE SLREQAEKEG SALSVRISNV.
The KIR (for inwardly rectifying potassium channel) family of potassium channels possess a greater tendency to allow potassium to flow into the cell rather than out of it. KIR4.1, also known as Kir1.2, is highly expressed in brain including glial cells, astrocytes and cortical neurons. KIR4.1 is also expressed in myelin-synthesizing oligodendrocytes and is crucial to myelination in the developing nervous system. The gene encoding human KIR4.1 maps to chromosome 1. KIR4.2, also known as Kir1.3, is expressed in kidney, lung, heart, thymus and thyroid during development. The gene encoding human KIR4.2 maps to chromosome 21 in the Down syndrome chromosome region 1, and KIR4.2 may play a role in the pathogenesis of Downs syndrome. KIR5.1 forms functional channels only by coexpression with either KIR4.1 or KIR4.2 in the kidney and pancreas.
Specifications
Kir4.1 (KCNJ10) | |
Polyclonal | |
Unconjugated | |
KCNJ10 | |
ATP-dependent inwardly rectifying potassium channel Kir4.1; ATP-sensitive inward rectifier potassium channel 10; ATP-sensitive inward rectifier potassium channel KAB-2; BIR10; BIRK1; BIRK-1; BIRK10; BIRK-10; brain-specific inwardly rectifying K(+) channel 1; glial ATP-dependent inwardly rectifying potassium channel KIR4.1; inward rectifier K(+) channel Kir1.2; Inward rectifier K(+) channel Kir4.1; inward rectifier K+ channel KIR1.2; Kab-2; Kcnj10; KCNJ13 PEN; KCNJ13-PEN; Kir1.2; Kir4.1; potassium channel, inwardly rectifying subfamily J member 10; potassium channel, inwardly rectifying subfamily J, member 10; potassium inwardly-rectifying channel, subfamily J, member 10; potassium voltage-gated channel subfamily J member 10; SESAME | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3766 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P78508 | |
KCNJ10 | |
Recombinant protein corresponding to Human KCNJ10. Recombinant protein control fragment (Product #RP-108298). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction