Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kir4.2 (KCNJ15) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575759
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575759 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575759 Supplier Invitrogen™ Supplier No. PA575759
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kir4.2 (KCNJ15)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Kcnj15
Gene Accession No. O88932, Q91ZF1, Q99712
Gene Alias 4930414N08Rik; AI182284; AI267127; ATP-sensitive inward rectifier potassium channel 15; Inward rectifier K(+) channel Kir1.3; inward rectifier K(+) channel Kir4.2; inward rectifier K+ channel KIR4.2; IRKK; KCNJ14; KCNJ15; KIR1.3; Kir4.2; Potassium channel, inwardly rectifying subfamily J member 15; potassium channel, inwardly rectifying subfamily J, member 15; potassium inwardly-rectifying channel, subfamily J, member 15; potassium voltage-gated channel subfamily J member 15
Gene Symbols Kcnj15
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human KCNJ15.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16516, 170847, 3772
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.