Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kir4.2 (KCNJ15) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550743
Description
The antibody detects endogenous levels of total KCNJ15 protein.
Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium.
Specifications
Kir4.2 (KCNJ15) | |
Polyclonal | |
Unconjugated | |
Kcnj15 | |
4930414N08Rik; AI182284; AI267127; ATP-sensitive inward rectifier potassium channel 15; Inward rectifier K(+) channel Kir1.3; inward rectifier K(+) channel Kir4.2; inward rectifier K+ channel KIR4.2; IRKK; KCNJ14; KCNJ15; KIR1.3; Kir4.2; Potassium channel, inwardly rectifying subfamily J member 15; potassium channel, inwardly rectifying subfamily J, member 15; potassium inwardly-rectifying channel, subfamily J, member 15; potassium voltage-gated channel subfamily J member 15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16516, 3772 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O88932, Q99712 | |
Kcnj15 | |
Synthetic peptide corresponding to a region derived from internal residues of human potassium inwardly-rectifying channel, subfamily J, member 15. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction